Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-2-(4-fluorophenyl)acetate 95%

Supplier: AMBEED, INC
Warning

A670511-1G A670511-250MG
76790-914EA 75 USD InStock
76790-914 76790-916
Methyl 2-bromo-2-(4-fluorophenyl)acetate 95%
Methyl 2-bromo-2-(4-fluorophenyl)acetate


Formula: C₉H₈BrFO₂
MDL Number: MFCD08444284
CAS Number: 71783-54-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty