Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetyl-4'-methylbiphenyl 98%

Supplier: AMBEED, INC
Warning

A671474-100MG A671474-1G A671474-250MG
76790-930EA 7 USD InStock
76790-930 76790-932 76790-934
4-Acetyl-4'-methylbiphenyl 98%
4-Acetyl-4'-methylbiphenyl
Precaución: for Research Use Only


Formula: C₁₅H₁₄O
MDL Number: MFCD00191374
CAS Number: 5748-38-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty