Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraisopropyl methylenebisphosphonate 98%

Supplier: AMBEED, INC
Warning

A687056-100G A687056-1G
76791-006EA 99 USD InStock
76791-006 76791-008
Tetraisopropyl methylenebisphosphonate 98%
Tetraisopropyl methylenebisphosphonate
Precaución: for Research Use Only


Formula: C₁₃H₃₀O₆P₂
MDL Number: MFCD00015021
CAS Number: 1660-95-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty