Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Suplatast tosylate 98%

Supplier: AMBEED, INC
Warning

A692262-10MG A692262-1MG A692262-50MG
76791-028EA 19 USD InStock
76791-028 76791-030 76791-032
Suplatast tosylate 98%
Suplatast tosylate
Precaución: for Research Use Only


Formula: C₂₃H₃₃NO₇S₂
MDL Number: MFCD00867604
CAS Number: 94055-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty