Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N,N-Trimethylanilinium tribromide 98%

Supplier: AMBEED, INC
Danger

A695524-10G A695524-1G A695524-25G A695524-5G
76791-034EA 9 USD InStock
76791-034 76791-036 76791-038 76791-040
N,N,N-Trimethylanilinium tribromide 98%
N,N,N-Trimethylanilinium tribromide
Precaución: for Research Use Only


Formula: C₉H₁₄Br₃N
MW: 375.93 g/mol
MDL Number: MFCD00011789
CAS Number: 4207-56-1
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty