Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptyl-p-toluenesulfonate 98%

Supplier: AMBEED, INC
Warning

A709151-10G A709151-25G A709151-5G
76791-078EA 24 USD InStock
76791-078 76791-080 76791-082
Heptyl-p-toluenesulfonate 98%
Heptyl-p-toluenesulfonate
Precaución: for Research Use Only


Formula: C₁₄H₂₂O₃S
MDL Number: MFCD00059442
CAS Number: 24767-82-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty