Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride 97%

Supplier: AMBEED, INC
Warning

A713210-100MG A713210-1G A713210-250MG
76791-104EA 19 USD InStock
76791-104 76791-106 76791-108
3-Amino-5-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride 97%
3-Amino-5-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₂₀BNO₄·ClH
MW: 313.585 g/mol
MDL Number: MFCD06659919
CAS Number: 850567-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty