Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc trifluoroacetate 98%

Supplier: AMBEED, INC
Warning

A723177-100G A723177-1G A723177-25G A723177-5G
76791-138EA 87 USD InStock
76791-138 76791-140 76791-142 76791-144
Zinc trifluoroacetate 98%
Zinc trifluoroacetate
Precaución: for Research Use Only


Formula: C₄F₆O₄Zn
MW: 291.43 g/mol
MDL Number: MFCD00061595
CAS Number: 21907-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty