Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triprolidine hydrochloride 99%

Supplier: AMBEED, INC
Warning

A728042-250MG A728042-25MG A728042-1G A728042-1MG
76791-188EA 42 USD InStock
76791-188 76791-190 77669-962 77669-964
Triprolidine hydrochloride 99%
Triprolidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₉H₂₅ClN₂O
MDL Number: MFCD00150574
CAS Number: 6138-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty