Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Undecanoyl chloride 98%

Supplier: AMBEED, INC
Danger

A729736-25G A729736-5G
76791-208EA 53 USD InStock
76791-208 77669-966
Undecanoyl chloride 98%
Undecanoyl chloride
Precaución: for Research Use Only


Formula: C₁₁H₂₁ClO
MW: 204.74 g/mol
MDL Number: MFCD00000739
CAS Number: 17746-05-3
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty