Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-β-Styrylboronic acid pinacol ester 96%

Supplier: AMBEED, INC
Warning

A746288-100MG A746288-1G A746288-250MG A746288-5G
76791-282EA 6 USD InStock
76791-282 76791-284 76791-286 76791-288
trans-β-Styrylboronic acid pinacol ester 96%
trans-beta-Styrylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₉BO₂
MDL Number: MFCD03453666
CAS Number: 83947-56-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty