Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TPPS hydrate (=tetraphenylporphyrin tetrasulfonic acid hydrate) 90%

Supplier: AMBEED, INC
Warning

A747070-100MG A747070-1G A747070-250MG
76791-304EA 72 USD InStock
76791-304 76791-306 77670-014
TPPS hydrate (=tetraphenylporphyrin tetrasulfonic acid hydrate) 90%
TPPS hydrate (=tetraphenylporphyrin tetrasulfonic acid hydrate)
Precaución: for Research Use Only


Formula: C₄₄H₃₀N₄O₁₂S₄
MW: 934.98 g/mol
MDL Number: MFCD00070632
CAS Number: 35218-75-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty