Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Xanthine sodium salt 98+%

Supplier: AMBEED, INC
Warning

A754150-1G A754150-250MG A754150-5G
76791-366EA 53 USD InStock
76791-366 76791-368 76791-370
Xanthine sodium salt 98+%
Xanthine sodium salt
Precaución: for Research Use Only


Formula: C₅H₃N₄NaO₂
MW: 174.095 g/mol
MDL Number: MFCD00069699
CAS Number: 1196-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty