Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-tert-Butyl-4-chlorobutyrophenone 96%

Supplier: AMBEED, INC
Warning

A758380-100G A758380-25G A758380-5G
76791-392EA 68 USD InStock
76791-392 76791-394 76791-396
4'-tert-Butyl-4-chlorobutyrophenone 96%
4'-tert-Butyl-4-chlorobutyrophenone
Precaución: for Research Use Only


Formula: C₁₄H₁₉ClO
MW: 238.7500 g/mol
MDL Number: MFCD00018996
CAS Number: 43076-61-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty