Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tiaprofenic acid 98%

Supplier: AMBEED, INC
Warning

A758829-100MG A758829-250MG
76791-398EA 67 USD InStock
76791-398 77670-042
Tiaprofenic acid 98%
Tiaprofenic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₃S
MW: 260.31 g/mol
MDL Number: MFCD00866089
CAS Number: 33005-95-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty