Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraethylammonium fluoride hydrate 97%

Supplier: AMBEED, INC
Warning

A767075-100G A767075-25G
76791-454EA 52 USD InStock
76791-454 76791-456 76791-458
Tetraethylammonium fluoride hydrate 97%
Tetraethylammonium fluoride hydrate
Precaución: for Research Use Only


Formula: C₈H₂₀FN·xH₂O
MW: 149.25 g/mol (anhy)
MDL Number: MFCD00149990
CAS Number: 98330-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty