Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-5-bromoindoline-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A770966-100MG A770966-1G A770966-250MG
76791-474EA 6 USD InStock
76791-474 76791-476 76791-478
tert-Butyl-5-bromoindoline-1-carboxylate 97%
tert-Butyl-5-bromoindoline-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₆BrNO₂
MW: 298.18 g/mol
MDL Number: MFCD08059280
CAS Number: 261732-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty