Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-1-ethyl-1H-pyrazole-4-carbonitrile 97%

Supplier: AMBEED, INC
Danger

A793572-100MG A793572-1G A793572-250MG
76791-598EA 85 USD InStock
76791-598 76791-600 76791-602
3-Amino-1-ethyl-1H-pyrazole-4-carbonitrile 97%
3-Amino-1-ethyl-1H-pyrazole-4-carbonitrile
Precaución: for Research Use Only


Formula: C₆H₈N₄
MDL Number: MFCD11040166
CAS Number: 122799-95-5
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty