Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A796238-1G A796238-250MG A796238-5G
76791-604EA 20 USD InStock
76791-604 76791-606 76791-608
6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester 97%
6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₆H₂₅BN₂O₄
MW: 320.196 g/mol
MDL Number: MFCD07781162
CAS Number: 910462-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty