Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-methoxybenzanilide 98%

Supplier: AMBEED, INC
Warning

A796709-100G A796709-25G A796709-5G
76791-610EA 50 USD InStock
76791-610 76791-612 76791-614 76791-616
3-Amino-4-methoxybenzanilide 98%
3-Amino-4-methoxybenzanilide
Precaución: for Research Use Only


Formula: C₁₄H₁₄N₂O₂
MW: 242.28 g/mol
MDL Number: MFCD00017166
CAS Number: 120-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty