Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsilyl acetate (acetoxytrimethylsilane) 98%

Supplier: AMBEED, INC
Danger

A805083-25G A805083-5G
76791-642EA 20 USD InStock
76791-642 76791-644
Trimethylsilyl acetate (acetoxytrimethylsilane) 98%
Trimethylsilyl acetate (acetoxytrimethylsilane)
Precaución: for Research Use Only


Formula: C₅H₁₂O₂Si
MW: 132.24 g/mol
MDL Number: MFCD00008695
CAS Number: 2754-27-0
UN: 3272
ADR: 3 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty