Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-3,7,11-Trimethyldodeca-1,6,10-trien-3-ol 95% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A808728-100G A808728-25G A808728-5G
76791-656EA 42 USD InStock
76791-656 76791-658 76791-660
(E)-3,7,11-Trimethyldodeca-1,6,10-trien-3-ol 95% stabilized
(E)-3,7,11-Trimethyldodeca-1,6,10-trien-3-ol
Precaución: for Research Use Only


Formula: C₁₅H₂₆O
MDL Number: MFCD00008911
CAS Number: 40716-66-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty