Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole 98%

Supplier: AMBEED, INC
Warning

A809890-100MG A809890-1G A809890-250MG
76791-666EA 8 USD InStock
76791-666 76791-668 76791-670
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole 98%
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole
Precaución: for Research Use Only


Formula: C₆H₈N₂
MDL Number: MFCD11870771
CAS Number: 59646-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty