Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraethylammonium trifluoroacetate 98%

Supplier: AMBEED, INC
Danger

A816376-5G A816376-25G
76791-690EA 11 USD InStock
76791-690 77670-154
Tetraethylammonium trifluoroacetate 98%
Tetraethylammonium trifluoroacetate
Precaución: for Research Use Only


Formula: C₁₀H₂₀F₃NO₂
MW: 243.26 g/mol
MDL Number: MFCD00192470
CAS Number: 30093-29-9
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty