Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Z-Dap(Z)-OH 97%

Supplier: AMBEED, INC
Warning

A829904-1G A829904-250MG A829904-5G
76791-782EA 18 USD InStock
76791-782 76791-784 76791-786
Z-Dap(Z)-OH 97%
Z-Dap(Z)-OH
Precaución: for Research Use Only


Formula: C₁₉H₂₀N₂O₆
MDL Number: MFCD00236966
CAS Number: 65621-26-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty