Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dansylglycine 98+%

Supplier: AMBEED, INC
Warning

A836722-100MG A836722-1G A836722-250MG A836722-5G
76791-832EA 26 USD InStock
76791-832 76791-834 76791-836 76791-838
Dansylglycine 98+%
Dansylglycine
Precaución: for Research Use Only


Formula: C₁₄H₁₆N₂O₄S
MDL Number: MFCD00037734
CAS Number: 1091-85-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty