Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Stiripentol 98%

Supplier: AMBEED, INC
Warning

A843564-10MG A843564-250MG A843564-25MG A843564-50MG A843564-5MG
76791-856EA 8 USD InStock
76791-856 76791-858 76791-860 76791-862 76791-864
Stiripentol 98%
Stiripentol
Precaución: for Research Use Only


Formula: C₁₄H₁₈O₃
MDL Number: MFCD00869310
CAS Number: 49763-96-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty