Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromo-4-fluoroaniline 96%

Supplier: AMBEED, INC
Warning

A881227-100MG A881227-1G A881227-250MG A881227-5G
76792-020EA 10 USD InStock
76792-020 76792-022 76792-024 76792-026
3,5-Dibromo-4-fluoroaniline 96%
3,5-Dibromo-4-fluoroaniline
Precaución: for Research Use Only


Formula: C₆H₄Br₂FN
MDL Number: MFCD09835195
CAS Number: 1003709-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty