Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propane-1,2,3-triyl triheptanoate ≥99%

Supplier: AMBEED, INC
Danger

A895125-1G A895125-25G A895125-5G A895125-100G
76792-044EA 11 USD InStock
76792-044 76792-046 76792-048 77670-308
Propane-1,2,3-triyl triheptanoate ≥99%
Propane-1,2,3-triyl triheptanoate
Precaución: for Research Use Only


Formula: C₂₄H₄₄O₆
MDL Number: MFCD00042910
CAS Number: 620-67-7
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty