Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-4-Phenylbut-3-enoic acid 95%

Supplier: AMBEED, INC
Warning

A939511-1G A939511-250MG A939511-25G A939511-5G
76792-160EA 25 USD InStock
76792-160 76792-162 76792-164 76792-166
(E)-4-Phenylbut-3-enoic acid 95%
(E)-4-Phenylbut-3-enoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MW: 162.188 g/mol
MDL Number: MFCD00002783
CAS Number: 1914-58-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty