Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1,2,3,4-tetrahydroisoquinoline-6-carboxylate hydrochloride 96%

Supplier: AMBEED, INC
Warning

A947077-100MG A947077-1G A947077-250MG
76792-186EA 26 USD InStock
76792-186 76792-188 76792-190
Methyl 1,2,3,4-tetrahydroisoquinoline-6-carboxylate hydrochloride 96%
Methyl 1,2,3,4-tetrahydroisoquinoline-6-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₄ClNO₂
MDL Number: MFCD03094737
CAS Number: 877861-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty