Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-779450 98+%

Supplier: AMBEED, INC
Danger

A986611-10MG A986611-1MG A986611-25MG A986611-50MG A986611-5MG
76792-316EA 28 USD InStock
76792-316 76792-318 76792-320 76792-322 76792-324
L-779450 98+%
L-779450
Precaución: for Research Use Only


Formula: C₂₀H₁₄ClN₃O
MDL Number: MFCD12828750
CAS Number: 303727-31-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty