Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-1-phenyl-2-phospholene-1-oxide 97%

Supplier: AMBEED, INC
Danger

A100603-1G A100603-25G A100603-5G
76796-230EA 12 USD InStock
76796-230 76796-232 76796-234
3-Methyl-1-phenyl-2-phospholene-1-oxide 97%
3-Methyl-1-phenyl-2-phospholene-1-oxide
Precaución: for Research Use Only


Formula: C₁₁H₁₃OP
MW: 192.2 g/mol
MDL Number: MFCD00014518
CAS Number: 707-61-9
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty