Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxynaphthalene-1-sulfonic acid 96%

Supplier: AMBEED, INC
Warning

A102559-100G A102559-10G A102559-1G A102559-25G A102559-500G A102559-5G
76796-322EA 43 USD InStock
76796-322 76796-324 76796-326 76796-328 76796-330 76796-332
5-Hydroxynaphthalene-1-sulfonic acid 96%
5-Hydroxynaphthalene-1-sulfonic acid
Precaución: for Research Use Only


Formula: C₁₀H₈O₄S
MDL Number: MFCD00065326
CAS Number: 117-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty