Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Erdosteine 98%

Supplier: AMBEED, INC
Warning

A103530-1G A103530-1MG A103530-250MG A103530-50MG A103530-5G
76796-374EA 40 USD InStock
76796-374 76796-376 76796-378 76796-380 76796-382
Erdosteine 98%
Erdosteine
Precaución: for Research Use Only


Formula: C₈H₁₁NO₄S₂
MDL Number: MFCD00867664
CAS Number: 84611-23-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty