Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ivabradine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A103870-100MG A103870-10MG A103870-1G A103870-250MG A103870-50MG
76796-392EA 18 USD InStock
76796-392 76796-394 76796-396 76796-398 76796-400
Ivabradine hydrochloride 98%
Ivabradine hydrochloride
Precaución: for Research Use Only


Formula: C₂₇H₃₆N₂O₅
MDL Number: MFCD00929899
CAS Number: 148849-67-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty