Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ibotenic acid 95%

Supplier: AMBEED, INC
Danger

A104575-10MG A104575-1MG A104575-25MG A104575-5MG
76796-418EA 289 USD InStock
76796-418 76796-420 76796-422 76796-424
Ibotenic acid 95%
Ibotenic acid
Precaución: For research use only.


Formula: C₅H₆N₂O₄
MDL Number: MFCD00069294
CAS Number: 2552-55-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty