Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diisopropyl malonate 95%

Supplier: AMBEED, INC
Warning

A107115-1000G A107115-100G A107115-10G A107115-25G A107115-500G
76796-586EA 26 USD InStock
76796-586 76796-588 76796-590 76796-592 76796-594
Diisopropyl malonate 95%
Diisopropyl malonate
Precaución: for Research Use Only


Formula: C₉H₁₆O₄
MW: 188.22 g/mol
MDL Number: MFCD00059359
CAS Number: 13195-64-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty