Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Riboflavine-5'-phosphate sodium salt 98% (by HPLC)

Supplier: AMBEED, INC
Warning

A107667-100G A107667-100MG A107667-10G A107667-1G A107667-25G A107667-25MG A107667-5G A107667-1MG A107667-500G
76796-612EA 212 USD InStock
76796-612 76796-614 76796-616 76796-618 76796-620 76796-622 76796-624 76796-626 77666-566 77666-568
Riboflavine-5'-phosphate sodium salt 98% (by HPLC)
Riboflavine-5'-phosphate sodium salt
Precaución: for Research Use Only


Formula: C₁₇H₂₀N₄NaO₉P
MW: 478.3256 g/mol
MDL Number: MFCD00065362
CAS Number: 130-40-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty