Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(hydroxymethyl)benzoate 98%

Supplier: AMBEED, INC
Warning

A108947-100G A108947-25G A108947-500G
76796-700EA 55 USD InStock
76796-700 76796-702 76796-704
Methyl-4-(hydroxymethyl)benzoate 98%
Methyl 4-(hydroxymethyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00009984
CAS Number: 6908-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty