Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfameter 98+%

Supplier: AMBEED, INC
Warning

A108984-1000G A108984-100G A108984-100MG A108984-25G A108984-500G
76796-706EA 165 USD InStock
76796-706 76796-708 76796-710 76796-712 76796-714
Sulfameter 98+%
Sulfameter
Precaución: for Research Use Only


Formula: C₁₁H₁₂N₄O₃S
MDL Number: MFCD00006067
CAS Number: 651-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty