Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Diethyl tartrate 98%

Supplier: AMBEED, INC
Warning

A109555-1000G A109555-100G A109555-10G A109555-25G A109555-500G
76796-734EA 53 USD InStock
76796-734 76796-736 76796-738 76796-740 76796-742 76796-744
L(+)-Diethyl tartrate 98%
L(+)-Diethyl tartrate
Precaución: for Research Use Only


Formula: C₈H₁₄O₆
MW: 206.2 g/mol
Boiling Pt: 280
Melting Pt: 18.7
MDL Number: MFCD00009143
CAS Number: 87-91-2
Merck Index: 13,03886
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty