Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amrinone 99

Supplier: AMBEED, INC
Danger

A110103-10G A110103-1MG A110103-250MG A110103-5G A110103-1G
76796-754EA 293 USD InStock
76796-754 76796-756 76796-758 76796-760 77666-602
Amrinone 99
Amrinone
Precaución: for Research Use Only


Formula: C₁₀H₉N₃O
MW: 187.202 g/mol
MDL Number: MFCD00083228
CAS Number: 60719-84-8
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty