Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Doripenem monohydrate 98%

Supplier: AMBEED, INC
Warning

A110224-100MG A110224-10MG A110224-1G A110224-1MG A110224-250MG A110224-50MG A110224-5G
76796-776EA 34 USD InStock
76796-776 76796-778 76796-780 76796-782 76796-784 76796-786 76796-788
Doripenem monohydrate 98%
Doripenem monohydrate
Precaución: for Research Use Only


Formula: C₁₅H₂₄N₄O₆S₂·H₂O
MDL Number: MFCD09749887
CAS Number: 364622-82-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty