Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Amino-5-(((benzyloxy)carbonyl)amino)pentanoic acid 95%

Supplier: AMBEED, INC
Warning

A110421-10G A110421-1G A110421-250MG A110421-5G
76796-796EA 71 USD InStock
76796-796 76796-798 76796-800 76796-802
(R)-2-Amino-5-(((benzyloxy)carbonyl)amino)pentanoic acid 95%
(R)-2-Amino-5-(((benzyloxy)carbonyl)amino)pentanoic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₈N₂O₄
MDL Number: MFCD00063115
CAS Number: 16937-91-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty