Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(Butylsulfonamido)-3-(4-hydroxyphenyl)propanoic acid 97%

Supplier: AMBEED, INC
Warning

A110832-10G A110832-1G A110832-250MG A110832-25G A110832-5G
76796-820EA 54 USD InStock
76796-820 76796-822 76796-824 76796-826 76796-828
(S)-2-(Butylsulfonamido)-3-(4-hydroxyphenyl)propanoic acid 97%
(S)-2-(Butylsulfonamido)-3-(4-hydroxyphenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₉NO₅S
MDL Number: MFCD07782139
CAS Number: 149490-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty