Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isatoic anhydride 98%

Supplier: AMBEED, INC
Warning

A111062-1000G A111062-100G A111062-25G A111062-500G
76796-842EA 31 USD InStock
76796-842 76796-844 76796-846 76796-848
Isatoic anhydride 98%
Isatoic anhydride
Precaución: for Research Use Only


Formula: C₈H₅NO₃
MDL Number: MFCD00006700
CAS Number: 118-48-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty