Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-(4-Benzylpiperidin-1-yl)-1-hydroxypropyl)phenol hemi((2R,3R)-2,3-dihydroxysuccinate) 98%

Supplier: AMBEED, INC
Warning

A114148-100MG A114148-10MG A114148-1MG A114148-50MG A114148-1G
76796-998EA 36 USD InStock
76796-998 76797-000 76797-002 76797-004 77666-658
4-(2-(4-Benzylpiperidin-1-yl)-1-hydroxypropyl)phenol hemi((2R,3R)-2,3-dihydroxysuccinate) 98%
4-(2-(4-Benzylpiperidin-1-yl)-1-hydroxypropyl)phenol hemi((2R,3R)-2,3-dihydroxysuccinate)
Precaución: for Research Use Only


Formula: C₄₆H₆₀N₂O₁₀
CAS Number: 23210-58-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty