Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6-amino-5-bromonicotinate 95%

Supplier: AMBEED, INC
Warning

A116099-100MG A116099-1G A116099-250MG A116099-25G A116099-5G
76797-064EA 12 USD InStock
76797-064 76797-066 76797-068 76797-070 76797-072
Ethyl 6-amino-5-bromonicotinate 95%
Ethyl 6-amino-5-bromonicotinate
Precaución: for Research Use Only


Formula: C₈H₉BrN₂O₂
MDL Number: MFCD06659894
CAS Number: 850429-51-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty