Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-5-oxo-D-prolinate 95%

Supplier: AMBEED, INC
Warning

A117790-100G A117790-1G A117790-25G A117790-5G
76797-172EA 145 USD InStock
76797-172 76797-174 76797-176 76797-178
tert-Butyl-5-oxo-D-prolinate 95%
tert-Butyl-5-oxo-D-prolinate
Precaución: for Research Use Only


Formula: C₉H₁₅NO₃
MDL Number: MFCD09261417
CAS Number: 205524-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty